Search
Search

Fam129c| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166213.1 | NP_001159685.1 | 10 - 11 | 1455 | 56 | |
| GenBank mRNA | AK158308.1 | - | 9 - 10 | 1426 | 56 | |
| AK163580.1 | - | 5 - 6 | 760 | 56 | ||
Set Membership: |
|