Search
Search

Ccdc107| Gene Symbol: Ccdc107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037913.2 | NP_001033002.1 | 2 - 3 | 325 | 91 | |
| GenBank mRNA | AK002915.1 | - | 2 - 3 | 293 | 91 | |
| AK004044.1 | - | 2 - 3 | 311 | 91 | ||
| BC027414.1 | - | 2 - 3 | 298 | 4236075 | 91 | |
Set Membership: |
|