| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10559457 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559458 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559480 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559483 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559462 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559485 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559474 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559456 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559459 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559472 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559481 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559484 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559464 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559478 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559467 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559454 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700019406.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019406.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019406.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019406.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019406.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019402.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019402.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019402.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019399.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019399.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019399.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019399.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019399.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019397.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019397.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019397.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019397.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019397.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019405.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019405.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019405.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019405.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019405.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019407.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019407.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019407.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019407.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019407.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019400.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019400.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019400.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019400.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019400.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019404.mm.1 |
NM_011090.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019404.mm.1 |
NM_011090.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019404.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019404.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019404.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019410.mm.1 |
NR_028115.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019410.mm.1 |
NR_028115.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019410.mm.1 |
TC0700004635.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1420464_s_at |
- |
- |
| Mouse Expression Set 430 |
1420464_PM_s_at |
- |
- |
| Murine Genome U74v2 Set |
100328_PM_s_at |
- |
- |
| Murine Genome U74v2 Set |
100328_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420464_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420464_PM_s_at |
- |
- |