| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2132832 |
NM_206868.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2132832 |
BC154405.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1987926 |
NM_001166758.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1987926 |
NM_001166714.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2132831 |
NM_206868.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2132831 |
BC154405.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49960 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
419392 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
423839 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435159 |
AK043858.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435159 |
AK079312.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435159 |
BC058996.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435159 |
NM_177102.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
965434 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
107379 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69312 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
251015 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
528109 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697102 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
195005 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
643408 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
336756 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
38121 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63001 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
571672 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308952 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
967230 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
213724 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
79200 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
840609 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
172619 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750516 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693649 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
595775 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
589331 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
102287 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
136735 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748653 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886244 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
338875 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187774 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746667 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
394471 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824165 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274636 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685917 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703104 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
264142 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354075 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
606777 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746436 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305700 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170714 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
107063 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
23860 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
755357 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
834549 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833036 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
847487 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
281395 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140694 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81849 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
69934 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
67003 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
144284 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111134 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
50971 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51746 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
41894 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
141539 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
123010 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1404 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97890 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
160494 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
119320 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128971 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
137097 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52183 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
141579 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
147372 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2132832 |
NM_206868.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2132832 |
BC154405.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2132831 |
NM_206868.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2132831 |
BC154405.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002834 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002837 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002828 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002821 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002864 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002841 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002822 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002857 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002860 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002865 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002844 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002847 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002816 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002839 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002859 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002846 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25002817 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135564 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05954290 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06524273 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05954280 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05954291 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05954277 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002870 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002806 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002880 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002864 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002855 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002883 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002803 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002877 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002886 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002816 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002859 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25002867 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471490 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001983 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002869 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002817 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002875 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06057791 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002872 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002834 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002876 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002871 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002859 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002839 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002805 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002811 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002841 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06220332 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06524266 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001980 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002880 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002802 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001978 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001984 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002874 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06220331 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06524254 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002828 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06441629 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002844 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001989 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002866 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002800 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06524256 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001987 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002857 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002831 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002826 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06230857 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002864 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002877 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002812 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06156672 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002824 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25001981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06220335 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06230860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002801 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05954285 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06220333 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002855 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06230861 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002803 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002820 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25002837 |
- |
- |