| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800000361.mm.1 |
NM_028175.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000361.mm.1 |
NM_028175.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000361.mm.1 |
TC0800000030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800000362.mm.1 |
NM_028175.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000362.mm.1 |
NM_028175.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000362.mm.1 |
TC0800000030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800000363.mm.1 |
NM_028175.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000363.mm.1 |
NM_028175.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000363.mm.1 |
TC0800000030.mm.1 |
MTA TC |