Search
Search

Haus7| Gene Symbol: Haus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028633.3 | NP_082909.1 | 1 - 2 | 189 | 98 | |
| NM_207104.1 | NP_996987.1 | 1 - 2 | 189 | 98 | ||
| GenBank mRNA | AK050729.1 | - | 1 - 2 | 123 | 98 | |
| AY029337.1 | - | 1 - 2 | 189 | 98 | ||
| BC025464.1 | - | 1 - 2 | 180 | 5351961 | 98 | |
Set Membership: |
|