| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10444818 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10444817 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1450582_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10444814 |
- |
- |
| Mouse Genome 430 2.0 Array |
1450582_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1450582_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700007036.mm.1 |
NR_051981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007036.mm.1 |
TC1700000685.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007040.mm.1 |
NR_051981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007040.mm.1 |
TC1700000685.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007038.mm.1 |
NR_051981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007038.mm.1 |
TC1700000685.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007039.mm.1 |
NR_051981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007039.mm.1 |
TC1700000685.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007037.mm.1 |
NR_051981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007037.mm.1 |
TC1700000685.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1450582_PM_at |
- |
- |
| Mouse Expression Set 430 |
1450582_at |
- |
- |
| Murine Genome U74v2 Set |
95773_PM_at |
- |
- |
| Murine Genome U74v2 Set |
95773_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1450582_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1450582_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1450582_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1450582_PM_at |
- |
- |