| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400032290.mm.1 |
TC0400003890.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032290.mm.1 |
NM_001017966.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032290.mm.1 |
NM_001017966.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032293.mm.1 |
TC0400003890.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032293.mm.1 |
NM_001017966.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032293.mm.1 |
NM_001017966.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032291.mm.1 |
TC0400003890.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032291.mm.1 |
NM_001017966.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032291.mm.1 |
NM_001017966.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032292.mm.1 |
TC0400003890.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032292.mm.1 |
NM_001017966.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032292.mm.1 |
NM_001017966.2 |
Refseq |