| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809003 |
AK015384.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809003 |
XM_001475855.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809003 |
XM_976954.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97660 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167554 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167555 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167552 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167570 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167569 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167551 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167556 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167560 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167549 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31167557 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P438781 |
BC139238.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P438781 |
BC139240.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P438781 |
NM_001011813.1 |
mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13335746 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13154874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167552 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167587 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167574 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167546 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167571 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167557 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167578 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167580 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167583 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167591 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167538 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167589 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167579 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167585 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167576 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167555 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167569 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167581 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167590 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167592 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31167540 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167560 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13335745 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167555 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167556 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167551 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167557 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167569 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167574 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13397759 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167573 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13335746 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167552 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167549 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13154873 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31167575 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13335748 |
- |
- |