| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10560371 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10560374 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10560372 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10560373 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10560370 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700021597.mm.1 |
TC0700002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021597.mm.1 |
NM_001029893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021597.mm.1 |
NM_001029893.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021598.mm.1 |
TC0700002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021598.mm.1 |
NM_001029893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021598.mm.1 |
NM_001029893.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021599.mm.1 |
TC0700002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021599.mm.1 |
NM_001029893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021599.mm.1 |
NM_001029893.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021600.mm.1 |
TC0700002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021600.mm.1 |
NM_001029893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021600.mm.1 |
NM_001029893.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021601.mm.1 |
TC0700002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021601.mm.1 |
NM_001029893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021601.mm.1 |
NM_001029893.1 |
Refseq |
| Murine Genome U74v2 Set |
168044_PM_at |
- |
- |
| Murine Genome U74v2 Set |
168044_at |
- |
- |