| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10394804 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10394803 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1453210_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10394802 |
- |
- |
| Mouse Genome 430 2.0 Array |
1453210_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453210_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1200001312.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001312.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001312.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001312.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001312.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001313.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001313.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001313.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001313.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001313.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001314.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001314.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001314.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001314.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001314.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001309.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001309.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001309.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001310.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001310.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001310.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001310.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001310.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001307.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001307.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001307.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001307.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001307.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001308.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001308.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001308.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001308.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001308.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001306.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001306.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001306.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001306.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001306.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001311.mm.1 |
NM_001033157.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001311.mm.1 |
NM_001033157.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001311.mm.1 |
NM_001201330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001311.mm.1 |
NM_001201330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001311.mm.1 |
TC1200000155.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1453210_at |
- |
- |
| Mouse Expression Set 430 |
1453210_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453210_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453210_at |
- |
- |