| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
AK132681.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
BC099567.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
AK005608.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
NR_027980.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
AK132681.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
BC099567.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
AK005608.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P515684 |
NR_027980.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2404484 |
AI605151.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2404484 |
BX513647.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2404484 |
NR_027980.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397669 |
AK036812.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397669 |
AK078407.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397669 |
AK078455.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397669 |
NM_023731.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897314 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469317 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520015 |
BC049631.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952460 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
222828 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
394901 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
686214 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
130736 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2404484 |
AI605151.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2404484 |
BX513647.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2404484 |
NR_027980.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P275125 |
AK132681.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P275125 |
BC099567.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P275125 |
AK005608.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P275125 |
NR_027980.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
AK132681.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
BC099567.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
AK005608.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
NR_027980.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
AK132681.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
BC099567.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
AK005608.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P515684 |
NR_027980.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P127818 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387943 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12050729 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387949 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12466710 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387915 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387928 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387930 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387953 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12271720 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387914 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387946 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387932 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387948 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12136852 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387938 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11845536 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387926 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387917 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387940 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387934 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387942 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30387944 |
- |
- |