Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03038854_s1
          See other 1600012H06RIK GE Assays ›
          Gene Symbol:
          1600012H06Rik
          Gene Name:
          RIKEN cDNA 1600012H06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 14943184 - 14945939 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03038854_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1600012H06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083880.1 NP_001077349.1 2 630 67
          NM_001083881.1 NP_001077350.1 2 700 67
          NM_001083882.1 NP_001077351.1 1 882 67
          NM_026451.2 NP_080727.2 2 635 67
          GenBank mRNA AK005427.1 - 2 612 67
          AK017276.1 - 1 812 67
          AK028052.1 - 2 636 67
          AK076083.1 - 2 592 67
          AK076441.1 - 839 67
          AK076476.1 - 2 555 67
          AK083064.1 - 1 823 67
          AK150772.1 - 2 655 67
          AK167490.1 - 2 642 67
          BC027313.1 - 2 614 3710655 67
          BC043303.1 - 1 843 67
          BC043449.1 - 1 843 67
          BC054359.1 - 2 632 67
          BC071250.1 - 2 547 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1600012H06Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062780 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062779 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC043449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC054359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC071250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC027313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC043303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC043449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC054359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC071250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC027313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P394385 BC043303.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199253 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162666 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC043449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC054359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC071250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC027313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC043303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC043449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC054359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC071250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC027313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P394385 BC043303.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31062796 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162666 - -
          Whole Mouse Genome G4122A A_52_P394385 BC043449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC054359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC071250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC027313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC043303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC043449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC054359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC071250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC027313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394385 BC043303.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108878 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162666 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13246934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31062780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31062791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13246941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31062787 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441920 - -
          Affymetrix HT MG-430 PM Array Plate 1428217_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428218_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448004 - -
          Affymetrix Mouse Gene 1.1 ST Array 10441919 - -
          Mouse Genome 430 2.0 Array 1428218_a_at - -
          Mouse Genome 430 2.0 Array 1428217_at - -
          Mouse Genome 430 2.0 Array 1428217_PM_at - -
          Mouse Genome 430 2.0 Array 1428218_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67912_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67912_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083880.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083880.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083882.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_001083882.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_026451.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 NM_026451.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001350.mm.1 TC1700000172.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001346.mm.1 NM_001083882.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001346.mm.1 NM_001083882.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001346.mm.1 TC1700000172.mm.1 MTA TC
          Mouse Expression Set 430 1428217_PM_at - -
          Mouse Expression Set 430 1428218_a_at - -
          Mouse Expression Set 430 1428218_PM_a_at - -
          Mouse Expression Set 430 1428217_at - -
          Murine Genome U74v2 Set 98891_PM_at - -
          Murine Genome U74v2 Set 98891_at - -
          Mouse Genome 430A 2.0 Array 1428217_PM_at - -
          Mouse Genome 430A 2.0 Array 1428218_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428217_at - -
          Mouse Genome 430A 2.0 Array 1428218_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428217_at - -
          Affymetrix HT Mouse Genome 430A Array 1428218_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428217_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428218_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919265 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3780707 - -
          MouseWG-6 v2 Expression BeadChip 6110603 - -
          MouseWG-6 v2 Expression BeadChip 6940592 - -
          Illumina mouse-6 v1.0 expression beadchip 6110603 - -
          Illumina mouse-6 v1.0 expression beadchip 3780707 - -
          Illumina mouse-6 v1.0 expression beadchip 6940592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2718861 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238211 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218419 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218419 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238211 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2718861 - -
          mouse-6 v1.1 (Illumina) scl067912.2_23-S - -
          mouse-6 v1.1 (Illumina) scl000097.1_10-S - -
          mouse-6 v1.1 (Illumina) scl000096.1_251-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780707 - -
          Illumina Sentrix mouse-6.v1 bead array 6940592 - -
          Illumina Sentrix mouse-6.v1 bead array 6110603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067912.2_23-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000097.1_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000096.1_251-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780707 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940592 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009738 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238211 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2718861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009738 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009735 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238211 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067912.2_23-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline