| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300014616.mm.1 |
NM_001001186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014616.mm.1 |
NM_001001186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014616.mm.1 |
TC1300002774.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300014615.mm.1 |
NM_001001186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014615.mm.1 |
NM_001001186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014615.mm.1 |
TC1300002774.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300018654.mm.1 |
NM_001001186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300018654.mm.1 |
NM_001001186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300018654.mm.1 |
TC1300002774.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300014614.mm.1 |
NM_001001186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014614.mm.1 |
NM_001001186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014614.mm.1 |
TC1300002774.mm.1 |
MTA TC |