Search
Search

Nol12| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133800.3 | NP_598561.2 | 1 - 2 | 145 | 144 | |
| GenBank mRNA | AK036891.1 | - | 1 - 2 | 88 | 144 | |
| AK037035.1 | - | 1 - 2 | 139 | 144 | ||
| AK083539.1 | - | 147 | 144 | |||
| AK167446.1 | - | 1 - 2 | 139 | 144 | ||
| AY940114.1 | - | 1 - 2 | 88 | 144 | ||
| BC094424.1 | - | 1 - 2 | 132 | 6819926 | 144 | |
Set Membership: |
|