| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700013624.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013624.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013624.mm.1 |
TC1700001540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013621.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013621.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013621.mm.1 |
TC1700001540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013619.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013619.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013619.mm.1 |
TC1700001540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013622.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013622.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013622.mm.1 |
TC1700001540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013623.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013623.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013623.mm.1 |
TC1700001540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013620.mm.1 |
NM_001104570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013620.mm.1 |
NM_001104570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013620.mm.1 |
TC1700001540.mm.1 |
MTA TC |