Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04210713_s1
          See other OLFR1323 GE Assays ›
          Gene Symbol:
          Olfr1323
          Gene Name:
          olfactory receptor 1323
          Gene Aliases:
          MOR216-1
          Chromosome Location:
          Chr.X: 50009306 - 50010235 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04210713_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1323
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146390.1 NP_666502.1 1 449 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1323

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972069 NM_146390.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114530 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972069 NM_146390.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165278 NM_146390.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32370958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114530 - -
          Whole Mouse Genome G4122A A_51_P165278 NM_146390.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123475 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15089629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14729010 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15089622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15089616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14729013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32370956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14962867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14859801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14859796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14817427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15089622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14729006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14938120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14859792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32370954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10604500 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604499 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2680164 - -
          Illumina mouse-6 v1.0 expression beadchip 2680164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714040 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714040 - -
          mouse-6 v1.1 (Illumina) scl54275.2.271_206-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54275.2.271_206-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714040 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0