Search
Search

Zfp398| Gene Symbol: Zfp398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027477.3 | NP_081753.1 | 4 - 5 | 904 | 98 | |
| NM_173034.3 | NP_766622.2 | 3 - 4 | 508 | 98 | ||
| GenBank mRNA | AK078250.1 | - | 3 - 4 | 475 | 98 | |
| AK081169.1 | - | 4 - 5 | 869 | 98 | ||
| AK089190.1 | - | 4 - 5 | 871 | 98 | ||
| AK155461.1 | - | 4 - 5 | 871 | 98 | ||
| BC112382.1 | - | 4 - 5 | 830 | 40059618 | 98 | |
Set Membership: |
|