Search
Search

Scarna10| Gene Symbol: Scarna10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028517.2 | 1 | 33 | 68 | ||
| GenBank mRNA | AF357346.1 | - | 1 | 33 | 68 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glyceraldehyde-3-phosphate dehydrogenase | Gapdh | Gapd | -19.9kb | Sense | Chr. 6 - 125161338 - 125166511 on NCBI Build GRCm38 |
| Downstream | mitochondrial ribosomal protein L51 | Mrpl51 |
2610511M02Rik, CDA09, HSPC241, Mrp64 |
5.8kb | Anti-sense | Chr. 6 - 125192200 - 125194392 on NCBI Build GRCm38 |
Set Membership: |
|