Search
Search

Nkx2-6| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010920.2 | NP_035050.2 | 1 - 2 | 249 | 76 | |
| GenBank mRNA | AF045150.1 | - | 1 - 2 | 249 | 76 | |
| BC100425.1 | - | 1 - 2 | 67 | 76 | ||
| BC132558.1 | - | 1 - 2 | 99 | 40130835 | 76 | |
| BC139035.1 | - | 1 - 2 | 99 | 8862057 | 76 | |
Set Membership: |
|