Search
Search

Gm1966| Gene Symbol: Gm1966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277179.1 | NP_001264108.1 | 3 | 7505 | 96 | |
| NM_001277180.1 | NP_001264109.1 | 3 | 7517 | 96 | ||
| NM_001277181.1 | NP_001264110.1 | 4 | 7589 | 96 | ||
| NM_001277182.1 | NP_001264111.1 | 4 | 7655 | 96 | ||
| NM_001277183.1 | NP_001264112.1 | 6 | 7789 | 96 | ||
Set Membership: |
|