Search
Search

Gm38425| Gene Symbol: Gm38425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251890.1 | NP_001238819.1 | 3 | 637 | 77 | |
| GenBank mRNA | DV646607.1 | - | 1 | 90 | 77 | |
| DV654665.1 | - | 3 | 637 | 77 | ||
Set Membership: |
|