Search
Search

4933402D24Rik| Gene Symbol: 4933402D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256158.1 | NP_001243087.1 | 2 - 3 | 242 | 116 | |
| GenBank mRNA | AK016612.1 | - | 2 - 3 | 243 | 116 | |
Set Membership: |
|