Search
Search

Trem1| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021406.5 | NP_067381.1 | 3 - 4 | 647 | 105 | |
| GenBank mRNA | AF241219.1 | - | 3 - 4 | 602 | 105 | |
| AK089439.1 | - | 3 - 4 | 616 | 105 | ||
| AK171753.1 | - | 3 - 4 | 616 | 105 | ||
| BC111875.1 | - | 2 - 3 | 259 | 105 | ||
Set Membership: |
|