Search
Search

1700001P01Rik| Gene Symbol: 1700001P01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028156.2 | NP_082432.2 | 1 - 2 | 273 | 97 | |
| GenBank mRNA | AK005619.1 | - | 1 - 2 | 237 | 97 | |
| BC048607.1 | - | 1 - 2 | 273 | 6741651 | 97 | |
| CA466212.1 | - | 241 | 97 | |||
Set Membership: |
|