Search
Search

Fam83f| Gene Symbol: Fam83f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145986.2 | NP_666098.2 | 4 - 5 | 1524 | 77 | |
| GenBank mRNA | AK030342.1 | - | 4 - 5 | 1506 | 77 | |
| AK145865.1 | - | 4 - 5 | 1524 | 77 | ||
| BC016600.1 | - | 4 - 5 | 1502 | 3153977 | 77 | |
Set Membership: |
|