Search
Search

Zc3h3| Gene Symbol: Zc3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172121.1 | NP_742119.1 | 1 - 2 | 58 | 62 | |
| GenBank mRNA | AJ516034.1 | - | 1 - 2 | 58 | 62 | |
| AK138281.1 | - | 1 - 2 | 66 | 62 | ||
| BC060682.1 | - | 1 - 2 | 57 | 6416246 | 62 | |
Set Membership: |
|