Search
Search

Csnka2ip| Gene Symbol: Csnka2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173861.2 | NP_776286.1 | 1 | 1219 | 67 | |
| GenBank mRNA | AF463503.1 | - | 1 | 1262 | 67 | |
| BC137818.1 | - | 1 | 1141 | 8860837 | 67 | |
Set Membership: |
|