Search
Search

4930544G11Rik| Gene Symbol: 4930544G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161773.1 | NP_001155245.1 | 1024 | 95 | ||
| GenBank mRNA | AK006363.1 | - | 1 | 984 | 95 | |
| AK016035.1 | - | 1 | 986 | 95 | ||
Set Membership: |
|