Search
Search

Fam46b| Gene Symbol: Fam46b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175307.6 | NP_780516.1 | 2 | 1422 | 85 | |
| GenBank mRNA | AK028944.1 | - | 2 | 1419 | 85 | |
| AK135706.1 | - | 2 | 1417 | 85 | ||
| AK137659.1 | - | 2 | 1423 | 85 | ||
| AK162145.1 | - | 2 | 1130 | 85 | ||
| AK163215.1 | - | 2 | 1422 | 85 | ||
| BC038001.1 | - | 2 | 1380 | 4913445 | 85 | |
Set Membership: |
|