Search
Search

SPINK5| Gene Symbol: SPINK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178719.3 | XP_012034109.1 | 24 - 25 | 2380 | 95 | |
| XM_012178720.3 | XP_012034110.1 | 24 - 25 | 2374 | 95 | ||
| XM_012178721.3 | XP_012034111.1 | 24 - 25 | 2377 | 95 | ||
| XM_012178722.3 | XP_012034112.1 | 24 - 25 | 2380 | 95 | ||
| XM_027970376.1 | XP_027826177.1 | 22 - 23 | 2170 | 95 | ||
Set Membership: |
|