Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012170103.3 | XP_012025493.1 | 4 - 5 | 445 | 147 | |
XM_012170108.3 | XP_012025498.1 | 4 - 5 | 379 | 147 | ||
XM_027966772.1 | XP_027822573.1 | 4 - 5 | 445 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |