Search
Search

LOC4340320| Gene Symbol: LOC4340320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785408.2 | XP_015640894.1 | 1 - 2 | 1034 | 60 | |
| GenBank mRNA | AK073624.1 | - | 1 - 2 | 433 | 60 | |
| AK101165.1 | - | 1 - 2 | 436 | 60 | ||
| AK102959.1 | - | 1 - 2 | 1038 | 60 | ||
Set Membership: |
|