Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195786.1 | NP_001182715.1 | 265 | 114 | ||
XM_015131728.2 | XP_014987214.2 | 430 | 114 | |||
XM_028830416.1 | XP_028686249.1 | 2238 | 114 | |||
GenBank mRNA | EG703544.1 | - | 245 | 114 | ||
JU336492.1 | - | 320 | 114 | |||
JV048307.1 | - | 299 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |