Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00471287_m1
          See other ATG3 GE Assays ›
          Gene Symbol:
          Atg3
          Gene Name:
          autophagy related 3
          Gene Aliases:
          Apg3l, PIG-1, Pig1
          Chromosome Location:
          Chr.11: 60585192 - 60613706 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00471287_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atg3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134394.2 NP_599221.1 2 - 3 401 92
          GenBank mRNA AF175224.1 - 2 - 3 404 92
          BC078743.1 - 2 - 3 413 7112937 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001573.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atg3

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          cellular protein modification process
          autophagy
          protein transport
          protein ubiquitination
          mitochondrial fragmentation involved in apoptotic process
          nucleophagy
          regulation of cilium assembly
          molecular_function
          Atg8 ligase activity
          Atg12 transferase activity
          ubiquitin-like protein transferase activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P667914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667914 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667914 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P667914 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P667914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68407 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753975 - -
          Affymetrix Rat Gene 1.0 ST Array 10753970 - -
          Affymetrix Rat Gene 1.0 ST Array 10753980 - -
          Affymetrix Rat Gene 1.0 ST Array 10753976 - -
          Affymetrix Rat Gene 1.0 ST Array 10753972 - -
          Affymetrix Rat Gene 1.0 ST Array 10753977 - -
          Affymetrix Rat Gene 1.0 ST Array 10753978 - -
          Affymetrix Rat Gene 1.0 ST Array 10753973 - -
          Affymetrix Rat Gene 1.0 ST Array 10753981 - -
          Affymetrix Rat Gene 1.0 ST Array 10753979 - -
          Affymetrix Rat Gene 1.0 ST Array 10753974 - -
          Affymetrix Rat Gene 1.1 ST Array 10753969 - -
          Rat Genome 230 2.0 Array 1398805_PM_at - -
          Rat Genome 230 2.0 Array 1398805_at - -
          Affymetrix HT Rat Focus Array 1398805_at - -
          Affymetrix HT Rat Focus Array 1398805_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100009578.rn.1 TC1100001573.rn.1 RTA TC
          Rat Expression Set 230 1398805_at - -
          Rat Expression Set 230 1398805_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753969 - -
          Rat Genome U34 Set rc_AI179248_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22058126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373265 - -
          RatRef-12 Expression BeadChip ILMN_64659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline