Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567566_m1
          See other PENK GE Assays ›
          Gene Symbol:
          Penk
          Gene Name:
          proenkephalin
          Gene Aliases:
          Enk, Penk-rs, Penk1, Penk2
          Chromosome Location:
          Chr.5: 17056412 - 17061762 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567566_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Penk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017139.1 NP_058835.1 1 - 2 466 56
          XM_006237835.1 XP_006237897.1 2 - 3 319 56
          GenBank mRNA BC083563.1 - 1 - 2 457 7112833 56
          M28263.1 - 2 - 3 187 56
          S49491.1 - 2 - 3 257 56
          Y07503.1 - 1 - 2 466 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002249.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Penk

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          startle response
          aggressive behavior
          neuropeptide signaling pathway
          locomotory behavior
          sensory perception of pain
          opioid peptide activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P749184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32452 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P749184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P749184 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P749184 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P749184 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P497807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875303 - -
          Affymetrix Rat Gene 1.0 ST Array 10875304 - -
          Affymetrix Rat Gene 1.0 ST Array 10875302 - -
          Affymetrix Rat Gene 1.0 ST Array 10875301 - -
          Affymetrix Rat Gene 1.1 ST Array 10875300 - -
          Rat Genome 230 2.0 Array 1367949_at - -
          Rat Genome 230 2.0 Array 1367949_PM_at - -
          Affymetrix HT Rat Focus Array 1367949_at - -
          Affymetrix HT Rat Focus Array 1367949_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500017107.rn.1 TC0500002249.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017108.rn.1 TC0500002249.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017109.rn.1 TC0500002249.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017106.rn.1 TC0500002249.rn.1 RTA TC
          Rat Expression Set 230 1367949_at - -
          Rat Expression Set 230 1367949_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875300 - -
          Rat Genome U34 Set S49491_s_at - -
          Rat Genome U34 Set rc_AI009300_at - -
          Rat Genome U34 Set rc_AI009300_at - -
          Rat Genome U34 Set S49491_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21480082 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352410 - -
          RatRef-12 Expression BeadChip ILMN_64242 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline