Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022536.2 | NP_071981.1 | 2 - 3 | 248 | 122 | |
GenBank mRNA | AF071225.1 | - | 2 - 3 | 248 | 122 | |
BC061971.1 | - | 2 - 3 | 277 | 5623342 | 122 | |
FQ211358.1 | - | 2 - 3 | 252 | 122 | ||
FQ212031.1 | - | 2 - 3 | 277 | 122 | ||
FQ219792.1 | - | 2 - 3 | 286 | 122 | ||
FQ219926.1 | - | 2 - 3 | 298 | 122 | ||
FQ221020.1 | - | 2 - 3 | 301 | 122 | ||
FQ221248.1 | - | 2 - 3 | 296 | 122 | ||
FQ221362.1 | - | 2 - 3 | 275 | 122 | ||
FQ222696.1 | - | 2 - 3 | 410 | 122 | ||
FQ223162.1 | - | 2 - 3 | 295 | 122 | ||
FQ228087.1 | - | 2 - 3 | 291 | 122 | ||
FQ229174.1 | - | 2 - 3 | 298 | 122 | ||
FQ229214.1 | - | 2 - 3 | 292 | 122 | ||
FQ230004.1 | - | 302 | 122 | |||
FQ231656.1 | - | 2 - 3 | 291 | 122 | ||
FQ232077.1 | - | 2 - 3 | 287 | 122 | ||
FQ232920.1 | - | 2 - 3 | 301 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Endogenous Controls Rattus norvegicus Rat Chemokine Panel Rat Endogenous Controls Inventoried Rat_Endogenous_Controls Probe spans exons |