Search
Search
| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031559.2 | NP_113747.2 | 13 - 14 | 1624 | 64 | |
| XM_006230695.1 | XP_006230757.1 | 14 - 15 | 1734 | 64 | ||
| GenBank mRNA | BC072522.1 | - | 13 - 14 | 1624 | 7105825 | 64 |
| L07736.1 | - | 13 - 14 | 1678 | 64 | ||
| U88294.1 | - | 12 - 13 | 1576 | 64 | ||
Set Membership: |
|