Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583505_m1
          See other STAT1 GE Assays ›
          Gene Symbol:
          Stat1
          Gene Name:
          signal transducer and activator of transcription 1
          Gene Aliases:
          DD6G4-4
          Chromosome Location:
          Chr.9: 54287540 - 54327958 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034164.1 NP_001029336.1 5 - 6 661 101
          NM_032612.3 NP_116001.2 5 - 6 661 101
          GenBank mRNA AF205604.1 - 3 - 4 371 101
          BC062079.1 - 5 - 6 661 5599645 101
          CV075250.1 - 5 - 6 579 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001944.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          JAK-STAT cascade
          response to nutrient
          blood circulation
          positive regulation of cell proliferation
          response to mechanical stimulus
          response to bacterium
          response to organic cyclic compound
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          cellular response to insulin stimulus
          tumor necrosis factor-mediated signaling pathway
          response to cytokine
          negative regulation of macrophage fusion
          cellular response to interferon-beta
          response to drug
          response to hydrogen peroxide
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          response to peptide hormone
          endothelial cell migration
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation by virus of viral protein levels in host cell
          positive regulation of smooth muscle cell proliferation
          response to cAMP
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          renal tubule development
          cellular response to lipopolysaccharide
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          tumor necrosis factor receptor binding
          enzyme binding
          CCR5 chemokine receptor binding
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein phosphatase 2A binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091988 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10593 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10593 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091988 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10593 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P164512 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10593 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12727 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700296892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57907 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39384 - -
          Agilent Rat cDNA Microarray (G4105A) 700296892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927843 - -
          Affymetrix Rat Gene 1.0 ST Array 10927868 - -
          Affymetrix Rat Gene 1.0 ST Array 10927864 - -
          Affymetrix Rat Gene 1.0 ST Array 10927853 - -
          Affymetrix Rat Gene 1.0 ST Array 10927859 - -
          Affymetrix Rat Gene 1.0 ST Array 10927867 - -
          Affymetrix Rat Gene 1.0 ST Array 10927865 - -
          Affymetrix Rat Gene 1.0 ST Array 10927857 - -
          Affymetrix Rat Gene 1.0 ST Array 10927851 - -
          Affymetrix Rat Gene 1.0 ST Array 10927860 - -
          Affymetrix Rat Gene 1.0 ST Array 10927850 - -
          Affymetrix Rat Gene 1.0 ST Array 10927845 - -
          Affymetrix Rat Gene 1.0 ST Array 10927848 - -
          Affymetrix Rat Gene 1.0 ST Array 10927866 - -
          Affymetrix Rat Gene 1.0 ST Array 10927858 - -
          Affymetrix Rat Gene 1.0 ST Array 10927870 - -
          Affymetrix Rat Gene 1.0 ST Array 10927849 - -
          Affymetrix Rat Gene 1.0 ST Array 10927846 - -
          Affymetrix Rat Gene 1.0 ST Array 10927854 - -
          Affymetrix Rat Gene 1.0 ST Array 10927863 - -
          Affymetrix Rat Gene 1.0 ST Array 10927852 - -
          Affymetrix Rat Gene 1.0 ST Array 10927844 - -
          Affymetrix Rat Gene 1.0 ST Array 10927855 - -
          Affymetrix Rat Gene 1.0 ST Array 10927862 - -
          Affymetrix Rat Gene 1.0 ST Array 10927861 - -
          Affymetrix Rat Gene 1.0 ST Array 10927847 - -
          Affymetrix Rat Gene 1.1 ST Array 10927842 - -
          Rat Genome 230 2.0 Array 1368835_at - -
          Rat Genome 230 2.0 Array 1384111_at - -
          Rat Genome 230 2.0 Array 1372757_PM_at - -
          Rat Genome 230 2.0 Array 1372757_at - -
          Rat Genome 230 2.0 Array 1387354_at - -
          Rat Genome 230 2.0 Array 1387354_PM_at - -
          Rat Genome 230 2.0 Array 1384111_PM_at - -
          Rat Genome 230 2.0 Array 1368835_PM_at - -
          Affymetrix HT Rat Focus Array 1387354_PM_at - -
          Affymetrix HT Rat Focus Array 1372757_PM_at - -
          Affymetrix HT Rat Focus Array 1372757_at - -
          Affymetrix HT Rat Focus Array 1387354_at - -
          Affymetrix Rat Clariom S Assay PSR0900015992.rn.1 TC0900001944.rn.1 RTA TC
          Rat Expression Set 230 1387354_at - -
          Rat Expression Set 230 1387354_PM_at - -
          Rat Expression Set 230 1372757_at - -
          Rat Expression Set 230 1368835_at - -
          Rat Expression Set 230 1372757_PM_at - -
          Rat Expression Set 230 1368835_PM_at - -
          Rat Expression Set 230 1384111_at - -
          Rat Expression Set 230 1384111_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927842 - -
          Rat Genome U34 Set rc_AA892553_at - -
          Rat Genome U34 Set AF055291mRNA_at - -
          Rat Genome U34 Set rc_AA999171_at - -
          Rat Genome U34 Set rc_AA849827_g_at - -
          Rat Genome U34 Set rc_AI105268_at - -
          Rat Genome U34 Set AF055291mRNA_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372159 - -
          RatRef-12 Expression BeadChip ILMN_69915 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline