Search
Search
| Gene Symbol: Mark2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021699.1 | NP_067731.1 | 3 - 4 | 321 | 122 | |
| XM_006230879.2 | XP_006230941.1 | 3 - 4 | 821 | 122 | ||
| XM_006230880.2 | XP_006230942.1 | 3 - 4 | 1608 | 122 | ||
| XM_006230881.2 | XP_006230943.1 | 3 - 4 | 340 | 122 | ||
| XM_006230883.2 | XP_006230945.1 | 3 - 4 | 820 | 122 | ||
| XM_006230884.2 | XP_006230946.1 | 3 - 4 | 819 | 122 | ||
| XM_006230885.2 | XP_006230947.1 | 3 - 4 | 819 | 122 | ||
| XM_006230887.2 | XP_006230949.1 | 3 - 4 | 818 | 122 | ||
| XM_008760169.1 | XP_008758391.1 | 3 - 4 | 820 | 122 | ||
| GenBank mRNA | Z83869.1 | - | 3 - 4 | 321 | 122 | |
Set Membership: |
|