Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013215.1 | NP_037347.1 | 1 - 2 | 271 | 115 | |
GenBank mRNA | AF045464.1 | - | 1 - 2 | 271 | 115 | |
BC078872.1 | - | 1 - 2 | 265 | 7127297 | 115 | |
BC089811.1 | - | 1 - 2 | 234 | 7126881 | 115 | |
X74673.1 | - | 1 - 2 | 268 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |