Search
Search
| Gene Symbol: Trak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133560.2 | NP_598244.2 | 2 - 3 | 466 | 63 | |
| XM_006244954.2 | XP_006245016.1 | 2 - 3 | 543 | 63 | ||
| XM_008767072.1 | XP_008765294.1 | 2 - 3 | 544 | 63 | ||
| XM_008767073.1 | XP_008765295.1 | 2 - 3 | 721 | 63 | ||
| GenBank mRNA | AF474163.1 | - | 1 - 2 | 96 | 63 | |
| AJ288898.2 | - | 2 - 3 | 466 | 63 | ||
| BC088393.1 | - | 2 - 3 | 465 | 7124967 | 63 | |
Set Membership: |
|