Search
Search
| Gene Symbol: Akt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017093.1 | NP_058789.1 | 3 - 4 | 461 | 138 | |
| XM_008759108.1 | XP_008757330.1 | 4 - 5 | 2860 | 138 | ||
| XM_008759109.1 | XP_008757331.1 | 3 - 4 | 448 | 138 | ||
| XM_008759110.1 | XP_008757332.1 | 3 - 4 | 2725 | 138 | ||
| XM_008759111.1 | XP_008757333.1 | 3 - 4 | 571 | 138 | ||
| GenBank mRNA | D30041.1 | - | 3 - 4 | 461 | 138 | |
| DQ198085.1 | - | 2 - 3 | 180 | 138 | ||
Set Membership: |
|