Search
Search

Slc10a2| Gene Symbol: Slc10a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017222.2 | NP_058918.1 | 2 - 3 | 600 | 148 | |
| XM_008771390.1 | XP_008769612.1 | 2 - 3 | 739 | 148 | ||
| GenBank mRNA | U07183.2 | - | 2 - 3 | 600 | 148 | |
Set Membership: |
|