Search
Search
| Gene Symbol: Ncstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174864.3 | NP_777353.1 | 5 - 6 | 676 | 68 | |
| XM_006250283.1 | XP_006250345.1 | 4 - 5 | 531 | 68 | ||
| XM_008769733.1 | XP_008767955.1 | 5 - 6 | 661 | 68 | ||
| XM_008769734.1 | XP_008767956.1 | 5 - 6 | 618 | 68 | ||
| XM_008769735.1 | XP_008767957.1 | 3 - 4 | 379 | 68 | ||
| GenBank mRNA | AF510722.1 | - | 4 - 5 | 460 | 68 | |
| AY101378.1 | - | 5 - 6 | 584 | 68 | ||
| BC074006.1 | - | 6 - 7 | 791 | 68 | ||
| BC167747.1 | - | 4 - 5 | 531 | 8365089 | 68 | |
Set Membership: |
|