Search
Search
| Gene Symbol: Arpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106919.1 | NP_001100389.1 | 7 - 8 | 662 | 62 | |
| XM_006245165.1 | XP_006245227.1 | 6 - 7 | 800 | 62 | ||
| XM_006245166.1 | XP_006245228.1 | 7 - 8 | 617 | 62 | ||
| GenBank mRNA | FQ214133.1 | - | 7 - 8 | 622 | 62 | |
| FQ226423.1 | - | 1 - 2 | 62 | 62 | ||
| FQ231947.1 | - | 4 - 5 | 415 | 62 | ||
Set Membership: |
|