Search
Search

LOC498122| Gene Symbol: LOC498122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109058.1 | NP_001102528.1 | 2 - 3 | 220 | 109 | |
| XM_008768900.1 | XP_008767122.1 | 1 - 2 | 524 | 109 | ||
Set Membership: |
|