Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prelid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271333.1 | NP_001258262.1 | 5 - 6 | 501 | 96 | |
XM_008772173.1 | XP_008770395.1 | 5 - 6 | 1147 | 96 | ||
XM_008772174.1 | XP_008770396.1 | 5 - 6 | 430 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |