Search
Search

Ankrd24| Gene Symbol: Ankrd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106771.1 | NP_001100241.1 | 10 - 11 | 1150 | 56 | |
| XM_008765093.1 | XP_008763315.1 | 10 - 11 | 1026 | 56 | ||
Set Membership: |
|