Search
Search
| Gene Symbol: Clasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769505.1 | XP_008767727.1 | 49 | 83 | ||
| XM_008769518.1 | XP_008767740.1 | 38 - 39 | 4582 | 83 | ||
| GenBank mRNA | BC087721.1 | - | 49 | 83 | ||
Set Membership: |
|